Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fluorescein 5(6)-isothiocyanate
Synonym(s):
FITC, Fluorescein isothiocyanate
Empirical Formula (Hill Notation):
C
21
H
11
NO
5
S
CAS Number:
27072-45-3
Molecular Weight:
389.38
EC Number:
248-207-7
Products
Product Category
dyes (1)
fluorescence reagent (1)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Physical Form
Product Line
Purity
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
46950
Fluorescein 5(6)-isothiocyanate,
BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Expand
View Pricing
F3651
Fluorescein 5(6)-isothiocyanate,
≥90% purity (HPLC), powder
Expand
View Pricing