Skip to Content
Merck
vibrant-m

FITC–DEAE–Dextran

vibrant-m
Synonym(s):
Fluorescein isothiocyanate–Diethylaminoethyl–Dextran

Products

Product Category
Brand
Biological Source
Physical Form
Chemical Composition
Conjugate
Quality Segment
Technique
Showing 1-3 of 3