Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Extractor body for Aldrich
®
Soxhlet extractor
Products
Brand
Sigma-Aldrich (4)
Capacity (mL)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z556238
Extractor body for Aldrich
®
Soxhlet extractor,
capacity 85 mL, top joint: ST/NS 45/50, bottom joint: ST/NS 24/40
Expand
View Pricing
Z556254
Extractor body for Aldrich
®
Soxhlet extractor,
capacity 200 mL, top joint: ST/NS 55/50, bottom joint: ST/NS 24/40
Expand
View Pricing
Z556211
Extractor body for Aldrich
®
Soxhlet extractor,
capacity 50 mL, top joint: ST/NS 34/45, bottom joint: ST/NS 24/40
Expand
View Pricing
Z556246
Extractor body for Aldrich
®
Soxhlet extractor,
capacity 145 mL, top joint: ST/NS 50/50, bottom joint: ST/NS 24/40
Expand
View Pricing