Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethyl benzoylacetate
Synonym(s):
Benzoylacetic acid ethyl ester, Ethyl 3-oxo-3-phenylpropionate, Ethyl benzoylacetate
Linear Formula:
C
6
H
5
COCH
2
COOC
2
H
5
CAS Number:
94-02-0
Molecular Weight:
192.21
EC Number:
202-295-3
Products
Product Category
building blocks (5)
Brand
Sigma-Aldrich (6)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technical (2)
Purum (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
281492
Ethyl benzoylacetate,
technical grade, 90%
Expand
View Pricing
689963
Ethyl benzoylacetate,
Arxada quality, ≥98% (GC)
Expand
View Pricing
12990
Ethyl benzoylacetate,
technical, ≥95% (HPLC)
Expand
View Pricing
8.01808
Ethyl benzoylacetate,
for synthesis
Expand
View Pricing
12980
Ethyl benzoylacetate,
redist., ≥97.0% (HPLC)
Expand
View Pricing
690082
Ethyl benzoylacetate,
Arxada quality, ≥91.0% (GC)
Expand
View Pricing