Skip to Content
Merck
Ethyl Alcohol 95% denatured, special

Ethanol (denatured)

Ethyl Alcohol 95% denatured, special
Synonym(s):
Ethanol denatured, Ethanol, Alcohol, Ethyl alcohol
Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Special Grade