
Esculin

Synonym(s):
6,7-Dihydroxycoumarin 6-glucoside, Aesculin, Bicolorin, Crataegin, Enallachrome, Escosyl, Esculetin 6-O-glucoside, Esculetin-6-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C15H16O9
CAS Number:
Molecular Weight:
340.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing