Skip to Content
Merck
Erythrosine B certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Erythrosine B

Erythrosine B certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Erythrosin extra bluish, 2′,4′,5′,7′-Tetraiodofluorescein disodium salt, Acid Red 51, Iodoeosin
Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86

Products