Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eriochrome
®
Black T
Synonym(s):
Mordant Black 11
Empirical Formula (Hill Notation):
C
20
H
12
N
3
NaO
7
S
CAS Number:
1787-61-7
Molecular Weight:
461.38
EC Number:
217-250-3
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Quality Segment
Application
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
858390
Eriochrome
®
Black T,
ACS reagent (indicator grade)
Expand
View Pricing
09-0400
Eriochrome
®
Black T,
SAJ special grade
Expand
View Pricing