Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Tubes 5.0 mL
Products
Product Category
tubes (7)
Color
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
EP0030122321
Eppendorf
®
Tubes 5.0 mL,
sterile, 5.0 mL, colorless, pkg of 200 tubes (2 bags of 100 tubes), with screw cap
Expand
View Pricing
EP0030122305
Eppendorf
®
Tubes 5.0 mL,
Eppendorf Quality, 5.0 mL, colorless, pkg of 200 tubes (2 bags of 100 tubes), with screw cap
Expand
View Pricing
EP0030119452
Eppendorf
®
Tubes 5.0 mL,
Eppendorf Quality, 5.0 mL, amber, pkg of 200 tubes (2 bags of 100 tubes), with hinged lid
Expand
View Pricing
EP0030122224
Eppendorf
®
Tubes 5.0 mL,
amber, sterile, pack of 200 ea
Expand
View Pricing
EP0030119606
Eppendorf
®
Tubes 5.0 mL,
Forensic DNA grade, 5.0 mL, colorless, pkg of 200 tubes (4 bags of 50 tubes), with hinged lid
Expand
View Pricing
EP0030122313
Eppendorf
®
Tubes 5.0 mL,
PCR clean, 5.0 mL, colorless, pkg of 200 tubes (2 bags of 100 tubes), with screw cap
Expand
View Pricing
EP0030122402
Eppendorf
®
Tubes 5.0 mL,
Forensic DNA grade, 5.0 mL, colorless, pkg of 200 tubes (4 bags of 50 tubes), with screw cap
Expand
View Pricing