Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Rotor Seal and Sealing Grease
Synonym(s):
Eppendorf
®
Rotor Seal and Sealing Grease
Products
Manufacturer
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP5495515003
Eppendorf
®
Rotor Seal and Sealing Grease,
for aerosol-tight rotor lid FA-24×2 (Centrifuge models 5420, 5425/R)
Expand
View Pricing
EP5409718002
Eppendorf
®
Rotor Seal and Sealing Grease,
for rotors FA-45-20-17 (Centrifuge models 5804/5804R/5810/5810R), and FA-20x5 (Centrifuge models 5920R, 5910R)
Expand
View Pricing
EP5495516000
Eppendorf
®
Rotor Seal and Sealing Grease,
for aerosol-tight rotor lid FA-10x5 and FA-18x2 (Centrifuge modle 5425/R)
Expand
View Pricing
EP5895177000
Eppendorf
®
Rotor Seal and Sealing Grease,
for rotor FA-6x250
Expand
View Pricing