Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Microplate 96/V-PP
Products
Product Category
plates (5)
Color
Manufacturer
Wells
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP0030603303
Eppendorf
®
Microplate 96/V-PP,
sterile (PCR clean), V-bottom clear wells, gray border, pkg of 80 plates (5 bags x 16 plates), DNA LoBind
Expand
View Pricing
MTP96VPWC-EP
Eppendorf
®
Microplate 96/V-PP,
sterile (PCR clean), V-bottom clear wells, white border, pkg of 80 plates (5 bags x 16 plates)
Expand
View Pricing
MTP96VPGW-EP
Eppendorf
®
Microplate 96/V-PP,
sterile (PCR clean), V-bottom white wells, gray border, pkg of 80 plates (5 bags x 16 plates)
Expand
View Pricing
MTP96VPWB-EP
Eppendorf
®
Microplate 96/V-PP,
sterile (PCR clean), V-bottom black wells, white border, pkg of 80 plates (5 bags x 16 plates)
Expand
View Pricing
EP951040227
Eppendorf
®
Microplate 96/V-PP,
sterile, white border, clear Wells, with conical bottom, pkg of 80 plates (5 bags x 16 plates)
Expand
View Pricing
EP951040243
Eppendorf
®
Microplate 96/V-PP,
sterile, V-bottom clear wells, white border, pack of 240 plates (10 bags × 24 plates)
Expand
View Pricing