Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean
Products
Product Category
pipette tips (16)
Manufacturer
Length (mm)
Volume (µL)
Color
Package Size
Size
Sterility
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z640549
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 500-2500 μL
Expand
View Pricing
EP022491717
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 0.1-20 μL, L 40 mm, medium gray, colorless tip, pack of 960 ea (10 trays × 96 ea)
Expand
View Pricing
Z640514
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 20-300 μL
Expand
View Pricing
Z640506
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 2-200 μL
Expand
View Pricing
EP022491733
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 0.1-200 μL, L 53 mm, yellow, colorless tip, pack of 960 ea (10 trays × 96 ea)
Expand
View Pricing
Z640522
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 50-1000 μL
Expand
View Pricing
EP022491750
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 50-1,000 μL, L 71 mm, blue, colorless tip, pack of 960 ea (10 trays × 96 ea)
Expand
View Pricing
Z640476
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 0.1-10 μL
Expand
View Pricing
ETIPRP20-EP
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume × L 0.5-20 μL × 46 mm, (light gray), pack of 960 ea (10 trays × 96 tips)
Expand
View Pricing
EP022491741
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 20-300 μL, L 55 mm, orange, colorless tip, pack of 960 ea (10 trays × 96 ea)
Expand
View Pricing
ETIPRP1250-EP
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume × L 50-1250 μL × 103 mm , L, (dark green), pack of 960 ea (10 racks × 96 tips)
Expand
View Pricing
Z640484
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 0.1-20 μL
Expand
View Pricing
EP022491776
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 0.25-2.5 mL, L 115 mm, red, colorless tip, pack of 480 ea (10 trays × 48 ea)
Expand
View Pricing
Z640530
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 50-1250 μL
Expand
View Pricing
EP022491709
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 0.1-10 μL, L 34 mm, dark gray, colorless tip, pack of 960 ea (10 trays × 96 ea)
Expand
View Pricing
EP022491768
Eppendorf
®
epT.I.P.S. Reloads, PCR Clean,
volume 50-1,250 μL, L 76 mm, green, colorless tip, pack of 960 ea (10 trays × 96 ea)
Expand
View Pricing