Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Adapter
Products
Product Category
adapters (5)
centrifuges (2)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
EP022639226
Eppendorf
®
Adapter,
to hold, 1 x 50mL, for 100 mL round bucket in Rotor A-4-38, pack of 2 ea
Expand
View Pricing
EP5920753002
Eppendorf
®
Adapter,
for 50 mL Oak Ridge tubes for rotor S-4xUniversal- Large
Expand
View Pricing
EP5910769008
Eppendorf
®
Adapter,
for 50 mL conical tubes, for rotor S-4xUniv., pkg of 2 pieces
Expand
View Pricing
EP5910764006
Eppendorf
®
Adapter,
for 50 mL tubes skirted bottom, for rotor S-4xUniv., pkg of 2 pieces
Expand
View Pricing
EP5910758006
Eppendorf
®
Adapter,
12 x 75 mm for S-4xUniversal, pack of 2 ea
Expand
View Pricing
EP5825739000
Eppendorf
®
Adapter,
for 14 x Tube 5.0 mL, for rotor S-4-104, set of 2 pieces
Expand
View Pricing
EP022639200
Eppendorf
®
Adapter,
to hold, 2 x 15mL, for 100 mL round bucket in Rotor A-4-38, pack of 2 ea
Expand
View Pricing