
Epoprostenol Related Compound A

Synonym(s):
6-Ketoprostaglandin F1α, (9α,11α,13E,15S)-9,11,15-Trihydroxy-6-oxoprosta-13-en-1-oic acid, 6-Oxo-prostaglandin F1α
Empirical Formula (Hill Notation):
C20H34O6
CAS Number:
Molecular Weight:
370.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing