
Epinastine for system suitability A

Synonym(s):
Epinastine hydrochloride, 9,13b-Dihydro-1H-dibenz[cf]imidazo[1,5-a]azepine hydrochloride, WAL-801Cl
Empirical Formula (Hill Notation):
C16H15N3 · HCl
CAS Number:
Molecular Weight:
285.77
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing