Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EPA 501/601 Trihalomethanes Calibration Mix
Products
Brand
Supelco (4)
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
certified reference material (4)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CRM48140
EPA 501/601 Trihalomethanes Calibration Mix,
certified reference material,
Trace
CERT
®
, 2000 μg/mL in methanol, ampule of 1 mL
Expand
View Pricing
CRM48746
EPA 501/601 Trihalomethanes Calibration Mix,
certified reference material,
Trace
CERT
®
, 200 μg/mL each component in methanol, ampule of 1 mL
Expand
View Pricing
CRM47904
EPA 501/601 Trihalomethanes Calibration Mix,
certified reference material,
Trace
CERT
®
, 100 μg/mL each component in methanol, ampule of 1 mL
Expand
View Pricing
4M8140-U
EPA 501/601 Trihalomethanes Calibration Mix,
certified reference material, 2000 μg/mL each component in methanol
Expand
View Pricing