Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ElectroGreen
®
Products
Brand
Sigma-Aldrich (9)
Sustainability
Color
Feature
Physical Form
Quality Segment
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
929654
ElectroGreen
®
,
Isopropyl Alcohol substitute for electronics, bio-based
Expand
View Pricing
929719
ElectroGreen
®
,
NMP substitute for electronics - type 1, bio-sourced
Expand
View Pricing
929662
ElectroGreen
®
,
NMP substitute for electronics – type 2, bio-sourced
Expand
View Pricing
929670
ElectroGreen
®
,
Acetone substitute for electronics, bio-sourced
Expand
View Pricing
929689
ElectroGreen
®
,
Toluene substitute for electronics, bio-sourced
Expand
View Pricing
929697
ElectroGreen
®
,
Butyl Cellosolve substitute for electronics, bio-sourced
Expand
View Pricing
929700
ElectroGreen
®
,
Xylene substitute for electronics, bio-sourced
Expand
View Pricing
929735
ElectroGreen
®
,
Cyclohexanone substitute for electronics, bio-sourced
Expand
View Pricing
929727
ElectroGreen
®
,
Methyl Amyl Ketone substitute for electronics, bio-sourced
Expand
View Pricing