Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Economical neoprene dry-box gloves
Products
Product Category
gloves (6)
Brand
Sigma-Aldrich (6)
Color
Manufacturer
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z170062
Economical neoprene dry-box gloves,
size 9, port size 8 in.
Expand
View Pricing
Z187313
Economical neoprene dry-box gloves,
size 8, port size 6 in.
Expand
View Pricing
Z187305
Economical neoprene dry-box gloves,
size 8, port size 8 in.
Expand
View Pricing
Z170070
Economical neoprene dry-box gloves,
size 10, port size 8 in.
Expand
View Pricing
Z187798
Economical neoprene dry-box gloves,
size 9, port size 6 in.
Expand
View Pricing
Z187801
Economical neoprene dry-box gloves,
size 10, port size 6 in.
Expand
View Pricing