Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
GL 45 connection system caps and accessories
Products
Product Category
caps (5)
Color
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z680397
Duran
®
GL 45 connection system caps and accessories,
GL 45 blue PP screw cap with 2 x GL 14 ports
Expand
View Pricing
Z680443
Duran
®
GL 45 connection system caps and accessories,
hose connection screw cap, blue PP GL 14
Expand
View Pricing
Z680494
Duran
®
GL 45 connection system caps and accessories,
insert for hose connection screw cap GL 14, 6.0 mm
Expand
View Pricing
Z680427
Duran
®
GL 45 connection system caps and accessories,
GL 45 blue PP screw cap with 4 ports for HPLC system
Expand
View Pricing
Z680419
Duran
®
GL 45 connection system caps and accessories,
GL 45 blue PP screw cap with 2 hose connections, 6-9 mm
Expand
View Pricing
Z680400
Duran
®
GL 45 connection system caps and accessories,
GL 45 blue PP screw cap with 3 x GL 14 ports
Expand
View Pricing
Z680435
Duran
®
GL 45 connection system caps and accessories,
spare part set for GL 14 ports on HPLC screw cap GL 45
Expand
View Pricing
Z680478
Duran
®
GL 45 connection system caps and accessories,
insert for hose connection screw cap GL 14, 3.0 mm
Expand
View Pricing
Z680486
Duran
®
GL 45 connection system caps and accessories,
insert for hose connection screw cap GL 14, 3.2 mm
Expand
View Pricing
Z680451
Duran
®
GL 45 connection system caps and accessories,
insert for hose connection screw cap GL 14, 1.6 mm
Expand
View Pricing
Z680508
Duran
®
GL 45 connection system caps and accessories,
spare membrane filter for pressure equalization set, 0.2 μm
Expand
View Pricing