Skip to Content
Merck
Duloxetine hydrochloride British Pharmacopoeia (BP) Reference Standard

Duloxetine hydrochloride

Duloxetine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
(S)-Duloxetine hydrochloride, (γS)-N-Methyl-γ-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride, (+)-(S)-N-Methyl-γ-(1-naphthyloxy)-2-thiophenepropylamine hydrochloride, (+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride, LY-248686 hydrochloride
Empirical Formula (Hill Notation):
C18H19NOS · HCl
CAS Number:
Molecular Weight:
333.88

Products

Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade