Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(+)-Muscarine chloride
Synonym(s):
(+)-(2S,4R,5S)-Tetrahydro-4-hydroxy-N,N,N,5-tetramethyl-2-furanmethanammonium chloride, (+)-Tetrahydro-4β-hydroxy-N,N,N,5α-tetramethyl-2α-furanmethanaminium chloride
Empirical Formula (Hill Notation):
C
9
H
20
ClNO
2
CAS Number:
2303-35-7
Molecular Weight:
209.71
EC Number:
218-963-2
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Physical Form
Purity
Research Area
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
M6532
(+)-Muscarine chloride,
~95% (TLC), powder
Expand
View Pricing