Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Distillation Standard
EC Number:
294-799-5
Products
Manufacturer
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CRMUDIGO
Distillation Standard,
ASTM D 86; IP 23 / ISO 3405 (168.5-360.1 °C)
Expand
View Pricing
CRMDIGO
Distillation Standard,
ASTM D 86; NF EN ISO 3405 (164-357 °C)
Expand
View Pricing
CRMDIGA
Distillation Standard,
ASTM D 86; NF EN ISO 3405 (30-178.6 °C)
Expand
View Pricing
CRMUDIKR
Distillation Standard,
ASTM D 86; NF EN ISO 3405 (148.1 - 260.3 °C)
Expand
View Pricing