Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Direct Thermal Cryo-Tags
®
Products
Product Category
labels (7)
Color
Manufacturer
Length (in)
Color
Technique
Filter Products
Showing 1-20 of 39
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z740246
Direct Thermal Cryo-Tags
®
,
Cryo-Spot, white, diam. 3/8 in.
Expand
View Pricing
Z740243
Direct Thermal Cryo-Tags
®
,
white, size 1.05 in. × 0.50 in.
Expand
View Pricing
Z740244
Direct Thermal Cryo-Tags
®
,
white, size 1.50 in. × 0.50 in.
Expand
View Pricing
Z743361
Direct Thermal Cryo-Tags
®
,
blue, L × W 1.50 in. × 0.75 in., pkg of 750 roll of
Expand
View Pricing
Z743395
Direct Thermal Cryo-Tags
®
,
white, L × W 2.0 in. × 0.75 in., pkg of 500 roll of
Expand
View Pricing
Z744112
Direct Thermal Cryo-Tags
®
,
white, W × L 1.69 in. × 0.75 in., pack of 500 ea (labels per rolllabels)
Expand
View Pricing
Z740247
Direct Thermal Cryo-Tags
®
,
white, size 2.0 in. × 0.25 in.
Expand
View Pricing
Z740245
Direct Thermal Cryo-Tags
®
,
white, size 1.50 in. × 0.75 in.
Expand
View Pricing
Z743359
Direct Thermal Cryo-Tags
®
,
blue, L × W 1.05 in. × 0.50 in., pkg of 1000 roll of
Expand
View Pricing
Z743377
Direct Thermal Cryo-Tags
®
,
green, L × W 1.05 in. × 0.50 in., pkg of 1000 roll of
Expand
View Pricing
Z743398
Direct Thermal Cryo-Tags
®
,
yellow, L × W 1.50 in. × 0.75 in., pkg of 750 roll of
Expand
View Pricing
Z743360
Direct Thermal Cryo-Tags
®
,
blue, L × W 1.50 in. × 0.50 in., pkg of 750 roll of
Expand
View Pricing
Z743388
Direct Thermal Cryo-Tags
®
,
red, L × W 1.50 in. × 0.75 in., pkg of 750 roll of
Expand
View Pricing
Z743382
Direct Thermal Cryo-Tags
®
,
orange, L × W 1.05 in. × 0.50 in., pkg of 1000 roll of
Expand
View Pricing
Z743362
Direct Thermal Cryo-Tags
®
,
blue, L × W 2.0 in. × 0.75 in., pkg of 500 roll of
Expand
View Pricing
Z743389
Direct Thermal Cryo-Tags
®
,
red, L × W 2.0 in. × 0.75 in., pkg of 500 roll of
Expand
View Pricing
Z743383
Direct Thermal Cryo-Tags
®
,
orange, L × W 1.50 in. × 0.50 in., pkg of 750 roll of
Expand
View Pricing
Z743386
Direct Thermal Cryo-Tags
®
,
red, L × W 1.05 in. × 0.50 in., pkg of 1000 roll of
Expand
View Pricing
Z743397
Direct Thermal Cryo-Tags
®
,
yellow, L × W 1.50 in. × 0.50 in., pkg of 750 roll of
Expand
View Pricing
Z743373
Direct Thermal Cryo-Tags
®
,
Cryo-Spot, white, diam. Type="regular">1/2 in., pkg of 4000 roll of
Expand
View Pricing
Page 1 of 2
Page 1 of 2