Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dichlorodimethylsilane
Synonym(s):
DMDCS, Dimethyldichlorosilane, Dichlorodimethylsilane
Linear Formula:
(CH
3
)
2
SiCl
2
CAS Number:
75-78-5
Molecular Weight:
129.06
EC Number:
200-901-0
Products
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
440272
Dichlorodimethylsilane,
≥99.5%
Expand
View Pricing
40140
Dichlorodimethylsilane,
≥98.5% (GC)
Expand
View Pricing
8.03452
Dichlorodimethylsilane,
for synthesis
Expand
View Pricing
80430
Dichlorodimethylsilane,
produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Expand
View Pricing