Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diallyl trisulfide
Synonym(s):
Allyl trisulfide, Di-2-propenyl trisulfide
Empirical Formula (Hill Notation):
C
6
H
10
S
3
CAS Number:
2050-87-5
Molecular Weight:
178.34
EC Number:
218-107-8
Products
Product Category
antioxidants (2)
inhibitors (2)
metabolites (2)
natural products (2)
sulfur compounds (2)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SMB00289
Diallyl trisulfide,
≥98% (HPLC)
Expand
View Pricing
SMB00577
Diallyl trisulfide,
≥97% (HPLC)
Expand
View Pricing