Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-Gluconic acid sodium salt
Synonym(s):
2,3,4,5,6-Pentahydroxycaproic acid sodium salt, D-Gluconate sodium salt, Sodium D-gluconate
Empirical Formula (Hill Notation):
C
6
H
11
NaO
7
CAS Number:
527-07-1
Molecular Weight:
218.14
EC Number:
208-407-7
Products
Product Category
carbohydrates (1)
monosaccharides (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Special Grade
certified reference material (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G9005
D-Gluconic acid sodium salt,
≥99% (HPLC)
Expand
View Pricing
64188
D-Gluconic acid sodium salt,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing