Skip to Content
Merck
Dextroamphetamine Related Compound A United States Pharmacopeia (USP) Reference Standard

Dextroamphetamine Related Compound A

Dextroamphetamine Related Compound A United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(±)-1-Phenyl-2-propanol, (±)-α-Methylphenethyl alcohol
Linear Formula:
C6H5CH2CH(OH)CH3
CAS Number:
Molecular Weight:
136.19

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1