Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dextran
Linear Formula:
[C
6
H
10
O
5
]
n
CAS Number:
9004-54-0
EC Number:
232-677-5
Products
Product Category
polysaccharides (10)
carbohydrates (2)
Brand
Sigma-Aldrich (1)
Supelco (11)
Physical Form
Format
Analyte Suitability
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (11)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
00271
Dextran,
analytical standard, 25,000
Expand
View Pricing
00269
Dextran,
analytical standard, 5,000
Expand
View Pricing
00268
Dextran,
analytical standard, 1,000
Expand
View Pricing
31394
Dextran,
enzymatic synth.
Expand
View Pricing
31430
Dextran,
analytical standard, suitable for gel permeation chromatography (GPC), Set M
p
1,000-400,000
Expand
View Pricing
00894
Dextran,
analytical standard, 270,000
Expand
View Pricing
00892
Dextran,
analytical standard, 80,000
Expand
View Pricing
00891
Dextran,
analytical standard, 50,000
Expand
View Pricing
00893
Dextran,
analytical standard, 150,000
Expand
View Pricing
00895
Dextran,
analytical standard, 410,000
Expand
View Pricing
00270
Dextran,
analytical standard, 12,000
Expand
View Pricing
00896
Dextran,
analytical standard, 670,000
Expand
View Pricing