Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dansyl chloride
Synonym(s):
5-(Dimethylamino)naphthalene-1-sulfonyl chloride, DNSCl, Dansyl Chloride
Empirical Formula (Hill Notation):
C
12
H
12
ClNO
2
S
CAS Number:
605-65-2
Molecular Weight:
269.75
EC Number:
210-092-6
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
D2625
Dansyl chloride,
BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Expand
View Pricing
39220
Dansyl chloride,
suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Expand
View Pricing
311155
Dansyl chloride,
Reagent used for fluorescent labeling of proteins, N-terminal amino acids, and amines.
Expand
View Pricing
03641
Dansyl chloride,
for HPLC derivatization, LiChropur
™
, ≥99.0% (HPLC)
Expand
View Pricing