Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CyDye
™
NIR-labelled secondary antibody
Products
Product Category
secondary antibodies (8)
Manufacturer
Research Area
Host
Species Reactivity
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GE29360784
CyDye
™
NIR-labelled secondary antibody,
700 nm, pkg of 0.1 mg, Cytiva 29360784
Expand
View Pricing
GE29360790
CyDye
™
NIR-labelled secondary antibody,
800 nm, pkg of 0.1 mg, Cytiva 29360790
Expand
View Pricing
GE29360786
CyDye
™
NIR-labelled secondary antibody,
700 nm, pkg of 0.1 mg, Cytiva 29360786
Expand
View Pricing
GE29360789
CyDye
™
NIR-labelled secondary antibody,
800 nm, pkg of 0.5 mg, Cytiva 29360789
Expand
View Pricing
GE29360791
CyDye
™
NIR-labelled secondary antibody,
800 nm, pkg of 0.5 mg, Cytiva 29360791
Expand
View Pricing
GE29360785
CyDye
™
NIR-labelled secondary antibody,
700 nm, pkg of 0.5 mg, Cytiva 29360785
Expand
View Pricing
GE29360787
CyDye
™
NIR-labelled secondary antibody,
700 nm, pkg of 0.5 mg, Cytiva 29360787
Expand
View Pricing
GE29360788
CyDye
™
NIR-labelled secondary antibody,
800 nm, pkg of 0.1 mg, Cytiva 29360788
Expand
View Pricing