
Creatinine hydrochloride

Synonym(s):
2-Amino-1-methyl-2-imidazolin-4-one hydrochloride
Empirical Formula (Hill Notation):
C4H7N3O · HCl
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing