Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
reusable low form beaker
Products
Product Category
beakers (17)
Manufacturer
Height (mm)
OD (mm)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
CLS1000P5L
Corning
®
reusable low form beaker,
polypropylene, size 5 L
Expand
View Pricing
CLS1000P400
Corning
®
reusable low form beaker,
polypropylene, size 400 mL
Expand
View Pricing
CLS1000P250
Corning
®
reusable low form beaker,
polypropylene, size 250 mL
Expand
View Pricing
CLS1000P100
Corning
®
reusable low form beaker,
polypropylene, size 100 mL
Expand
View Pricing
CLS1000P1L
Corning
®
reusable low form beaker,
size 1 L
Expand
View Pricing
CLS1000P2L
Corning
®
reusable low form beaker,
polypropylene, size 2 L
Expand
View Pricing
CLS1000P150
Corning
®
reusable low form beaker,
polypropylene, size 150 mL
Expand
View Pricing
CLS1000P600
Corning
®
reusable low form beaker,
polypropylene, size 600 mL
Expand
View Pricing
CLS1000P50
Corning
®
reusable low form beaker,
polypropylene, size 50 mL
Expand
View Pricing
CLS1000P3L
Corning
®
reusable low form beaker,
polypropylene, size 3 L
Expand
View Pricing
CLS1003P1L
Corning
®
reusable low form beaker,
perfluoroalkoxy copolymer, size 1 L
Expand
View Pricing
CLS1000P25
Corning
®
reusable low form beaker,
polypropylene, size 25 mL
Expand
View Pricing
CLS1003P500
Corning
®
reusable low form beaker,
perfluoroalkoxy copolymer, size 500 mL
Expand
View Pricing
CLS1003P250
Corning
®
reusable low form beaker,
perfluoroalkoxy copolymer, size 250 mL
Expand
View Pricing
CLS1003P25
Corning
®
reusable low form beaker,
perfluoroalkoxy copolymer, size 25 mL
Expand
View Pricing
CLS1003P50
Corning
®
reusable low form beaker,
perfluoroalkoxy copolymer, size 50 mL
Expand
View Pricing
CLS1003P100
Corning
®
reusable low form beaker,
perfluoroalkoxy copolymer, size 100 mL
Expand
View Pricing