Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
narrow mouth Erlenmeyer flask, reusable
Products
Product Category
flasks (10)
Manufacturer
Capacity (L)
Capacity (mL)
Cap
Package Size
Sterility
Style
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
CLS4985P100
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 100 mL, polypropylene, with GL-40 PP screw cap
Expand
View Pricing
CLS4985P50
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 50 mL, polypropylene, with GL-40 PP screw cap
Expand
View Pricing
CLS4985P250
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 250 mL, polypropylene, with GL-52 PP screw cap
Expand
View Pricing
CLS4985P1L
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 1 L, polypropylene, with GL-52 PP screw cap
Expand
View Pricing
CLS4990P50
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 50 mL, polymethylpentene, with GL-40 PP screw cap
Expand
View Pricing
CLS4985P500
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 500 mL, polypropylene, with GL-52 PP screw cap
Expand
View Pricing
CLS4990P250
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 250 mL, polymethylpentene, with GL-52 PP screw cap
Expand
View Pricing
CLS4990P100
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 100 mL, polymethylpentene, with GL-40 PP screw cap
Expand
View Pricing
CLS4990P1L
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 1 L, polymethylpentene, with GL-52 PP screw cap
Expand
View Pricing
CLS4990P500
Corning
®
narrow mouth Erlenmeyer flask, reusable,
capacity 500 mL, polymethylpentene, with GL-52 PP screw cap
Expand
View Pricing