Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Filtr
EX
™
96 well filter plates
Products
Product Category
plates (4)
Color
Manufacturer
Binding Type
Wells
Color
Membrane Material
Plate Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS3504
Corning
®
Filtr
EX
™
96 well filter plates,
0.2 μm pore size non-treated surface white polystyrene plate, PVDF membrane (hydrophilic), round wells, round bottom white polystyrene , rectangular bottom white polystyrene wells non-sterile, case of 50, pack of 10
Expand
View Pricing
CLS3511
Corning
®
Filtr
EX
™
96 well filter plates,
0.66 mm, non-treated surface white polystyrene plate, round white polystyrene , rectangular bottom white polystyrene wells, glass fiber membrane, non-sterile, case of 50, pack of 10
Expand
View Pricing
CLS3505
Corning
®
Filtr
EX
™
96 well filter plates,
0.2 μm pore size non-treated surface white polystyrene plate, round white polystyrene , round wells, rectangular bottom white polystyrene wells, PVDF membrane, sterile, case of 50, pack of 10, pkg of 50 ea
Expand
View Pricing
CLS3510
Corning
®
Filtr
EX
™
96 well filter plates,
0.25 mm, non-treated surface white polystyrene plate, round white polystyrene , rectangular bottom white wells, glass fiber membrane, non-sterile, case of 50, pack of 10
Expand
View Pricing