Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Falcon
®
Cell Culture Dish
Synonym(s):
Cell Culture Dish
Products
Product Category
dishes (6)
Feature
Manufacturer
Binding Type
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS353002
Corning
®
Falcon
®
Cell Culture Dish,
plate diam. × capacity 60 x 15 mm × 6 mL, Tissue Culture (TC)-treated surface, sterile
Expand
View Pricing
CLS353025
Corning
®
Falcon
®
Cell Culture Dish,
plate diam. × capacity 150 x 25 mm × 45 mL, with 20mm Grid Molded in Base, Tissue Culture (TC)-treated surface, sterile
Expand
View Pricing
CLS353001
Corning
®
Falcon
®
Cell Culture Dish,
plate diam. × capacity 35 x 10 mm × 2.5 mL, Tissue Culture (TC)-treated surface, sterile
Expand
View Pricing
CLS353003
Corning
®
Falcon
®
Cell Culture Dish,
plate diam. × capacity 100 x 20 mm × 16 mL, Tissue Culture (TC)-treated surface, sterile
Expand
View Pricing
CLS353004
Corning
®
Falcon
®
Cell Culture Dish,
plate diam. × capacity 60 x 15 mm × 6 mL, Tissue Culture (TC)-treated surface, sterile
Expand
View Pricing
CLS353037
Corning
®
Falcon
®
Cell Culture Dish,
plate diam. × capacity 60 x 15 mm × 6 mL, Tissue Culture (TC)-treated surface, sterile
Expand
View Pricing