Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
BioCoat
™
Poly-D-Lysine Microplate
Products
Product Category
plates (21)
Manufacturer
Binding Type
Wells
Color
Sterility
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CLS354640
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear black wells, lid
Expand
View Pricing
CLS354461
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear wells, lid
Expand
View Pricing
CLS356661
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, flat bottom white wells, lid
Expand
View Pricing
CLS356660
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, flat bottom clear white wells, lid
Expand
View Pricing
CLS354660
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, flat bottom clear white wells, lid
Expand
View Pricing
CLS356651
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear white wells, lid
Expand
View Pricing
CLS356640
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear black wells
Expand
View Pricing
CLS356620
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom opaque white wells, lid
Expand
View Pricing
CLS356690
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear wells
Expand
View Pricing
CLS354661
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, flat bottom white wells, lid
Expand
View Pricing
CLS356697
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, clear black wells
Expand
View Pricing
CLS354651
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear white wells, lid
Expand
View Pricing
CLS354620
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom opaque white wells, lid
Expand
View Pricing
CLS354663
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, flat bottom clear black wells, lid
Expand
View Pricing
CLS356461
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear wells, lid
Expand
View Pricing
CLS356663
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, flat bottom clear wells, lid
Expand
View Pricing
CLS356693
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear wells
Expand
View Pricing
CLS356692
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom clear black wells
Expand
View Pricing
CLS354662
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 384 wells, flat bottom clear wells, lid
Expand
View Pricing
CLS356691
Corning
®
BioCoat
™
Poly-D-Lysine Microplate,
well size 96 wells, flat bottom white wells, lid
Expand
View Pricing
Page 1 of 2
Page 1 of 2