Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper/nickel
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
GF55981659
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, size 95 × 95 mm
Expand
View Pricing
GF07217119
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, length 2 m, coil width 192 mm
Expand
View Pricing
GF93263407
Copper/nickel,
rod, 12.7 mm diameter, Cu70%/Ni30%, length 100 mm, temper as drawn
Expand
View Pricing
GF28140791
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, 4 mm diameter
Expand
View Pricing
GF08379007
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, length 0.5 m, coil width 192 mm
Expand
View Pricing
GF60527268
Copper/nickel,
foil, Cu70%/Ni30%, thickness 0.016 mm, size 190 × 190 mm
Expand
View Pricing
GF93535409
Copper/nickel,
foil, thickness 0.016 mm, Cu70%/Ni30%, diameter 50 mm
Expand
View Pricing
GF02055608
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, 8 mm diameter
Expand
View Pricing
GF18951127
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, 25 mm diameter
Expand
View Pricing
GF82101248
Copper/nickel,
foil, Cu70%/Ni30%, thickness 0.016 mm, length 0.2 m, coil width 192 mm
Expand
View Pricing
GF53165943
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, length 1 m, coil width 192 mm
Expand
View Pricing
GF19270650
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, 10 mm diameter
Expand
View Pricing
GF42629249
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, 15 mm diameter
Expand
View Pricing
GF03744682
Copper/nickel,
foil, Cu 70%/Ni 30%, thickness 0.016 mm, 6 mm diameter
Expand
View Pricing