
cis-Aconitic acid

Synonym(s):
cis-Propene-1,2,3-tricarboxylic acid
Linear Formula:
HO2CCH2C(CO2H)=CHCO2H
CAS Number:
Molecular Weight:
174.11
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing