
Choloylglycine Hydrolase from Clostridium perfringens (C. welchii)

Synonym(s):
3α,7α,12α-Trihydroxy-5β-cholan-24-oylglycine amidohydrolase
CAS Number:
Products
Brand
Physical Form
Research Area
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing