Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chlorhexidine Hydrochloride
Synonym(s):
Chlorhexidine dihydrochloride, 1,6-Bis(
N
5
-[
p
-chlorophenyl]-
N
1
-biguanido)hexane; 1,1′-Hexamethylenebis(5-[
p
-chlorophenyl]biguanide), Lisium
Empirical Formula (Hill Notation):
C
22
H
30
Cl
2
N
10
· 2HCl
CAS Number:
3697-42-5
Molecular Weight:
578.37
EC Number:
223-026-6
Products
Brand
Supelco (1)
Formula Weight
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
PHR1459
Chlorhexidine Hydrochloride,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing