Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chlorazol Black
Synonym(s):
4-Amino-3-[[4′-[(2,4-diaminophenyl)azo][1,1′-biphenyl]-4-yl]azo]-5-hydroxy-6-(phenylazo)-2,7-naphthalenedisulfonic acid disodium salt, Chlorazole Black E, Direct Black 38
Empirical Formula (Hill Notation):
C
34
H
25
N
9
Na
2
O
7
S
2
CAS Number:
1937-37-7
Molecular Weight:
781.73
EC Number:
217-710-3
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C1144
Chlorazol Black,
Powder
Expand
View Pricing