Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cesium iodide
Empirical Formula (Hill Notation):
CsI
CAS Number:
7789-17-5
Molecular Weight:
259.81
EC Number:
232-145-2
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Special Grade
Anhydrous (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
203033
Cesium iodide,
99.999% trace metals basis
Expand
View Pricing
914819
Cesium iodide,
AnhydroBeads
™
, 99.999% trace metals basis, (Perovskite grade)
Expand
View Pricing
202134
Cesium iodide,
99.9% trace metals basis
Expand
View Pricing
429384
Cesium iodide,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing
21004
Cesium iodide,
analytical standard, suitable for mass spectrometry (MS)
Expand
View Pricing