Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cesium fluoride
Synonym(s):
NSC 84270
Empirical Formula (Hill Notation):
CsF
CAS Number:
13400-13-0
Molecular Weight:
151.90
EC Number:
236-487-3
Products
Product Category
salts (5)
Brand
Sigma-Aldrich (5)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
198323
Cesium fluoride,
99%
Expand
View Pricing
289345
Cesium fluoride,
99.9% trace metals basis
Expand
View Pricing
20990
Cesium fluoride,
purum p.a., ≥98.0%
Expand
View Pricing
255718
Cesium fluoride,
99.99% trace metals basis
Expand
View Pricing
20989
Cesium fluoride,
BioUltra, ≥99.0% (F)
Expand
View Pricing