Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Centrifuge tube with screw cap, conical bottom, co-polymer
Products
Product Category
tubes (6)
Color
Manufacturer
Agency
Capacity (mL)
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
AXYSCT15ML25S
Centrifuge tube with screw cap, conical bottom, co-polymer,
capacity 15 mL, sterile
Expand
View Pricing
AXYSCT15ML500
Centrifuge tube with screw cap, conical bottom, co-polymer,
capacity 15 mL
Expand
View Pricing
AXYSCT50ML25S
Centrifuge tube with screw cap, conical bottom, co-polymer,
capacity 50 mL, sterile
Expand
View Pricing
AXYSCT50ML500
Centrifuge tube with screw cap, conical bottom, co-polymer,
capacity 50 mL, non-sterile
Expand
View Pricing
AXYSCT15MLRS
Centrifuge tube with screw cap, conical bottom, co-polymer,
capacity 15 mL, units of 25, sterile
Expand
View Pricing
AXYSCT50MLRS
Centrifuge tube with screw cap, conical bottom, co-polymer,
capacity 50 mL, sterile
Expand
View Pricing