
CellPrime® r LR3 IGF powder

Synonym(s):
CellPrime® R3 IGF-I human
Empirical Formula (Hill Notation):
C400H619N111O115S9
CAS Number:
Molecular Weight:
9111.45
Products
Brand
Formula Weight
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing