Skip to Content
Merck
Cathepsin D from bovine spleen lyophilized powder, ≥2.0 units/mg protein

Cathepsin D from bovine spleen

Cathepsin D from bovine spleen lyophilized powder, ≥2.0 units/mg protein
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Manufacturer
Physical Form
Tissue
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing