
Caspofungin diacetate

Synonym(s):
1-[(4R,5S)-5-[(2-Aminoethyl)amino]-N2-(10,12-dimethyl-1-oxotetradecyl)-4-hydroxy-L-ornithine]-5-[(3R)-3-hydroxy-L-ornithine]-pneumocandin B0
Empirical Formula (Hill Notation):
C52H88N10O15 · 2C2H4O2
CAS Number:
Molecular Weight:
1213.42
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing