Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbopack
™
B
Synonym(s):
Carbopack
™
B
Products
Product Category
tubes (3)
Brand
Supelco (3)
Sustainability
Feature
Product Line
Agency
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
29754-U
Carbopack
™
B,
O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Expand
View Pricing
30136-U
Carbopack
™
B,
glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
™
Expand
View Pricing
29535-U
Carbopack
™
B,
glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Expand
View Pricing