Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cameo
™
Filtration Membrane
Synonym(s):
Cameo
™
Filtration Membrane
Products
Product Category
syringe filters (4)
Manufacturer
Diameter (mm)
Membrane Material
Pore size (µm)
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z741970
Cameo
™
Filtration Membrane,
polycarbonate membrane (Track Etched (PCTE)), pore size 0.4 μm, diam. 47 mm, pkg of 100 ea
Expand
View Pricing
Z741974
Cameo
™
Filtration Membrane,
Nylon membrane, pore size 5.0 μm, diam. 47 mm, pkg of 100 ea
Expand
View Pricing
Z741971
Cameo
™
Filtration Membrane,
nitrocellulose membrane, pore size 0.45 μm, diam. 47 mm, pkg of 100 ea
Expand
View Pricing
Z741972
Cameo
™
Filtration Membrane,
Nylon membrane, pore size 0.45 μm, diam. 47 mm, pkg of 100 ea
Expand
View Pricing