Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium carbide
Linear Formula:
CaC
2
CAS Number:
75-20-7
Molecular Weight:
64.10
EC Number:
200-848-3
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Purity
Special Grade
Technical (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
21039
Calcium carbide,
granulated, technical, ≥75% (gas-volumetric)
Expand
View Pricing
270296
Calcium carbide,
pieces, thickness <10 mm , typically, technical grade, ~80%
Expand
View Pricing